missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SI Analytics™ TitroLine™ 7800
SI Analytics™ TitroLine™ 7800 enhances the universal features of the TitroLine™ 7750 with an additional IDS measurement input.
Brand: SI Analytics™ 285221030
8417.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- pH/mV-input with 24 bit transducer Electrode socket according to DIN 19 262 or additional with BNC socket insert RFID receiver for SI Analytics ID electrodes
- Valve: PTFE/ETFE
- Cylinder: borosilicate glass 3.3 (DURAN™)
- Hoses: FEP, blue
- Highly visible full color display, that can be easily viewed from a distance and extreme angles
- With new interchangeable modules which all relevant reagent and unit data can be stored
- Expandable thanks to the 2 x USB-host, 1 x USB-PC, 1 LAN and 2 x RS232 ports. Connectable are e.g. USB keyboard, USB printer, barcode reader, USB flash drives, balances, PC und further SI Analytics devices such as piston burettes and sample changers
- Storage of results using via USB port (PDF – and CSV -format) including method transfer
- With standard methods for potentiometric and KF titration
- Second digital measuring port for intelligent digital sensors (IDS)
Specifications
0.1/0.1 = ± 1 Digital | |
Two measuring inputs, magnetic stirrer, and TitriSoft 3.2 | |
TitroLine™ 7800 | |
10°C to 40°C |
3.5 inches -1/4 VGA TFT display with 320x240 pixels | |
Polypropylene | |
-40°C to +125°C | |
29.6 x 15.3 x 45 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction