Learn More
SI Analytics TitroLine 7800 Basic Unit
Robust, reliable, ready-to-use, all-rounder for potentiometric titration with an additional digital measuring input.
Brand:  SI Analytics 285220980
Product Code. 15554923
  • 6505.00 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Can also be used as volumetric KF titrator
  • High visible full colour display, that can be easily viewed from a distance and extreme angles
  • With intelligent interchangeable modules and storage of all relevant reagent data
  • Includes two USB-A, one USB-B, two RS-232 and one LAN port for expansion and connection of devices such as keyboard, printer, barcode reader, memory sticks, balances, PC an further SI Analytics devices
  • Storage of results via USB port (PDF and CSV -format) including method transfer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Storage of results via USB port (PDF and CSV -format) including method transfer
Benchtop Titrator
Includes two USB-A, one USB-B, two RS-232 and one LAN port for expansion and connection of devices such as keyboard, printer, barcode reader, memory sticks, balances, PC an further SI Analytics devices.
TitroLine 7800 Basic Unit