Learn More
Velp Scientifica™ TITROLINE 5000 Automatic Titrator
Velp Scientifica™ TITROLINE 5000 Automatic Titrator with compact new titrator for simple routine titrations.
Brand:  Velp Scientifica™ R30800194
Product Code. 15349494
  • 7725.00 € / Each
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Titrations to pH, mV - endpoint (2 EP)
  • Titrations to one equivalence point (pH und mV) with dynamic or linear titration addition
  • Manual titrations with the controller
  • Dosing tasks
  • Number of methods: 3
  • Salt content in foodstuffs (cheese, soy sauce, ketchup)
  • Total acidity in wine and beverages
  • Nitrogen according to Kjeldahl
  • Chloride in dringing water and other samples
  • Alkalinity (p + m-value)
  • FOS/TAC
Specifications
Specifications
3.5 in. color touch screen
Combined distillation and titration, for simultaneous Kjeldahl nitrogen/protein determination
416 mm
Titroline Easy K
Technopolymer