Learn More
Tuttnauer Tiva8 Glass Washer Disinfector
Product Code. 17960050 Shop All Tuttnauer Products
Click to view available options
Description:
Glass Door
Glass Door, Lab Configuration, Racks and Nozzles
Stainless Steel Door
Stainless Steel Door, No Drying
Wide with Glass Door and Side Cabinet
Wide with Glass Door and Side Cabinet, No Drying
Unit Size:
1 unit
Product Code. 17960050
Brand: Tuttnauer TIVA8XE1ML
Only null left
Add to basket
Add to basket
The Tiva8 Glass Washer Disinfector provides a flexible turnkey solution for washing, disinfecting and drying diverse laboratory equipment for a complete decontamination life cycle. The washer can be loaded up to 4 washing levels with a large variety of washing trolleys and accessories.
Small, reliable and customizable. The Tiva 8 washer disinfector provides a flexible turnkey solution for washing, disinfecting and drying diverse laboratory equipment for a complete decontamination life cycle. The TIVA 8 washer can be loaded up to 4 washing levels with a large variety of washing trolleys and accessories.
  • Diverse load types
    • Available trolleys for DIN baskets, containers, mini-invasive equipment, anesthesia kits, bottles and shoes
  • Washing chamber made of high quality AISI 316L
    • 165 Liter chamber, perfect for private clinics
  • Manual glass door
    • Hinged, manual door
  • Advanced Touch Control Touch panel with color graphic display with a total of 40 programs: preset and programmable procedures
Advanced features
  • Air drying prefilter and HEPA H14 filter.
  • 3 heating elements.
  • Built-in steam condenser.
  • Integrated water softener (optional).
  • Dosing pumps with flowmeter
  • For acid and alkaline detergents
  • 2 additional pumps optional
Specifications
Description Stainless Steel Door, No Drying
Display LCD
Material Stainless Steel
Product Type Laboratory Glassware Washer
Volume (English) 43.6 gal.
Volume (Metric) 165 L