Learn More
Cole-Parmer™ Thin Layer Chromatography (TLC) Plate Holder, 25 Plate Capacity
Plastic construction resists common TLC solvents.
Brand:  Cole-Parmer™ 34105-60
123.50 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Product Code. 12379389
  • 130.00 € / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Hold up to 25 TLC plates of any size. Bottom grooves slope toward back vertical support grooves.
Specifications
Specifications
Thin Layer Chromatography (TLC) Plate Holder, 25 Plate Capacity
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Thin Layer Chromatography (TLC) Plate Holder, 25 Plate Capacity >