Learn More
Macherey-Nagel™ TLC Simultaneous Developing Chamber
Develop multiple TLC plates simultaneously. TLC Simultaneous Developing Chambers come with a choice of room for either 2 or 5 plates.
Brand:  Macherey-Nagel™ 814018
 View more versions of this product
Product Code. 12374613
  • 228.00 € / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Develop multiple TLC plates simultaneously. TLC Simultaneous Developing Chambers come with a choice of room for either 2 or 5 plates.
  • Accessory for TLC
  • Chambers are thick walled
  • Clear glass tank 
  • Heavy ground glass lid
Specifications
Specifications
1 Pk.
TLC Simultaneous Developing Chamber
For 2 Plates 10 x 10 cm
2