missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM110 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA562501
This item is not returnable.
View return policy
Description
Immunogen sequence: VDNFLMRKGK TKAKLEERGA NQDSRNGSKV RYRRAASHEE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Acts as a regulator of store-operated Ca(2+) entry (SOCE) at junctional sites that connect the endoplasmic reticulum (ER) and plasma membrane (PM), called ER-plasma membrane (ER-PM) junction or cortical ER (PubMed:26322679, PubMed:26644574). SOCE is a Ca(2+) influx following depletion of intracellular Ca(2+) stores (PubMed:26322679). Acts by interacting with STIM1, promoting STIM1 conformational switch (PubMed:26322679). Involved in STIM1 relocalization to ER-PM junctions (PubMed:26644574). Contributes to the maintenance and reorganization of store-dependent ER-PM junctions (PubMed:26644574). [UniProt]Specifications
TMEM110 | |
Polyclonal | |
Unconjugated | |
STIMATE | |
1810038N08Rik; 2310014H19Rik; AW554125; STIM-activating enhancer; STIM-activating enhancer encoded by TMEM110; STIMATE; Store-operated calcium entry regulator STIMATE; Tmem110; Transmembrane protein 110 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375346 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86TL2 | |
STIMATE | |
Recombinant protein corresponding to Human TMEM110 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only