missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMT™ Elution Buffer and Anti-TMT Resin

For enhanced enrichment, use Thermo Scientific TMT Elution Buffer with Immobilized Anti-TMT Resin for peptides that have been chemically labeled with Tandem Mass Tag (TMT) Isobaric Label Reagents for multiplexed protein quantitation using tandem mass spectrometry.

108.00 € - 768.00 €

Specifications

Product Line TMT™
Label or Dye Tandem Mass Tag™ (TMT) Labels
Starting Material Cell Lysate
Detection Method Mass Spectrometry
Workflow Step Protein Labeling
View More Specs

Products 2
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
13444289
View Documents
Thermo Scientific™
90104
TMT™ Elution Buffer 20 mL
108.00 €
20mL
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
 
11532591
View Documents
Thermo Scientific™
90076
Immobilized Anti-TMT™ Antibody Resin 6 mL
768.00 €
6mL
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
 
Description

Description

Tandem mass tag (TMT) isobaric label reagents are used for multiplexed protein quantitation using tandem mass spectrometry. Immobilized Anti-TMT Resin is designed to effectively capture and enrich peptides or proteins labeled using any TMT reagent. Anti-TMT Resin and TMT Elution Buffer are effective for reducing sample complexity, improving dynamic range, and studying cysteine modifications such as S-nitrosylation, oxidation, and disulfide bonds of protein samples labeled with iodoTMT reagents.

Tandem mass tag (TMT) reagents are used to chemically label proteins in different samples for multiplexed quantitation using tandem mass spectrometry. Immobilized Anti-TMT Resin contains an agarose-immobilized anti-TMT monoclonal antibody and is effective for capture and enrichment of peptides or proteins labeled using any TMT reagent. This helps simplify otherwise complex samples before LC-MS/MS analysis, especially when the targets for labeling are rare (e.g., cysteine sulfhydryls). Enrichment of labeled peptides by this method results in fewer peptides identified per protein, but can significantly increase the number of unique proteins identified, making it easier to detect and measure low-abundance proteins.

Immobilized Anti-TMT Resin uses our highly specific Anti-TMT Antibody (Cat. No. 90075) to bind peptides labeled with TMT reagents. The antibody is specific for the mass reporter region of the TMT reagents, which allows for enrichment of TMT-labeled peptides regardless of peptide labeling chemistry (e.g., amine-reactive, sulfhydryl-reactive). TMT Elution Buffer is a volatile neutral buffer that competitively elutes captured TMT-labeled peptides. When used in combination with Immobilized Anti-TMT Resin, TMT Elution Buffer provides higher specific enrichment of TMT-labeled peptides compared to acidic or basic elution buffers.

Enrichment features include:
Specific—uses specific, high-affinity antibody for the TMT structure (Kd >100 pM)
Compatible—effective for all varieties of TMT reagents (amine-reactive or cysteine-reactive)
Versatile—use resin with TMT Elution Buffer for optimal sample enrichment

Specifications

Specifications

TMT™
Cell Lysate
Protein Labeling
Tandem Mass Tag™ (TMT) Labels
Mass Spectrometry
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • TMT Elution Buffer

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : triethylamine, acetic acid

SDS
Documents

Documents

Product Certifications