Learn More
Thermo Scientific™ TMT11-131C Label Reagent
Tandem mass tag (TMT) reagents are specially designed to enable identification and quantitation of proteins in different samples using tandem mass spectrometry.
351.00 € - 6090.00 €
Specifications
Product Type Isobaric Label Reagent
Detection Method Mass Spectrometry
View More Specs
Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15706663
View Documents
Thermo Scientific™
A34807
1 x 5 mg
532.00 €
5mg
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
15877944
View Documents
Thermo Scientific™
A37724
3 x 0.8 mg
351.00 €
2.40mg
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
15722695
View Documents
Thermo Scientific™
A34808
1 x 5 mg (per tag)
6090.00 €
5mg
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
15988062
View Documents
Thermo Scientific™
A37725
3 x 0.8 mg (per tag)
3505.00 €
Pack of 3
In Stock
to see stock.
Add to basket
Description
Description
Tandem mass tag (TMT) reagents are specially designed to enable identification and quantitation of proteins in different samples using tandem mass spectrometry. The TMT11-131C tag structure is identical to the other TMT10pex tags, but the TMT11-131C mass reporter is labeled with only 13C isotopes. The addition of the TMT11-131C tag enables separation of the 131-mass reporter ion of the original TMT10plex reagents into N and C variants, creating an 11th channel for relative quantitation using high resolution Orbitrap MS instruments.
The advantages of TMT label reagents include increased multiplexed relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. TMT10plex label reagents plus TMT11-131C is ideal for analysis of multiple protein samples from inhibitor dose response experiments, time course experiments, or to improve statistics on biological replicates.
Features of this TMT11-131C Label Reagent include:
Multiplex—concurrent MS analysis of up to 11 samples derived from cells, tissues, or biological fluids
Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
Efficient—amine-reactive, NHS-ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
Compatible—optimized for use with high resolution Thermo Scientific Orbirtrap MS/MS platforms, with data analysis fully supported by Proteome Discoverer 2.3
TMT11-131C is available as a standalone tag (Cat. No. A34807, A37724), as part of this TMT10plex Isobaric Label Reagent Set plus TMT11-131C, or as part of the complete 11plex set of reagents in a 96-well plate format. When combined with the industry-leading, high resolution Orbitrap instruments and software, TMT reagents provide integrated total solutions for quantitative protein expression analysis.
Specifications
Specifications
Isobaric Label Reagent
Mass Spectrometry