missing translation for 'onlineSavingsMsg'
Learn More

TNC/Tenascin-C Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen TNC/Tenascin-C
Concentration 450 μg/mL
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17211396
View Documents Promotion Details
Proteintech
27789-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
17201396
View Documents Promotion Details
Proteintech
27789-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The TNC/Tenascin-C antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TNC/Tenascin-C. This antibody recognizes human antigen. The TNC/Tenascin-C antibody has been validated for the following applications: WB,ELISA analysis.

Specifications

Specifications

TNC/Tenascin-C
Western Blot, ELISA
Unconjugated
Rabbit
3371
450 μg/mL
Polyclonal
Liquid
Human
Primary
Videos
SDS
Documents

Documents

Product Certifications