missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TNF beta ProQuantum Immunoassay Kit
GREENER_CHOICE

ELISA

788.00 € - 3530.00 €

Specifications

Accession Number P01374
Assay Range 0.064 to 25000 pg/mL
Assay Sensitivity <0.05 pg/mL
Conjugate FAM™
Detection Method Taqman™ Real-time PCR
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16381994
GREENER_CHOICE
View Documents
Invitrogen™
A42898
96 Tests
788.00 €
Pack of 96
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
 
17114729
GREENER_CHOICE
View Documents
Invitrogen™
A428985
5 x 96 Tests
3530.00 €
Each
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
 
Description

Description

TNF beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. The protein is highly inducible, secreted, and forms heterotrimers with lymphotoxin-beta which anchor lymphotoxin-alpha to the cell surface. This protein also mediates a large variety of inflammatory, immunostimulatory, and antiviral responses, is involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis. Genetic variations in this gene are associated with susceptibility to leprosy type 4 and psoriatic arthritis.
Specifications

Specifications

P01374
<0.05 pg/mL
Taqman™ Real-time PCR
ProQuantum
Human
qPCR Instruments
1 hr.
FAM™
Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
TNF beta
2 hrs
0.064 to 25000 pg/mL
FAM™
Proximity-based Immunoassay
Plasma, Serum, Supernatant
Protein Quantitation
4049
Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
RUO
Frozen at -20°C
30 mins
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.