Learn More
TNFR1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A6E3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 60192-1-IG-150UL
 View more versions of this product
Product Code. 16817185
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
TNFR1 (Tumor necrosis factor receptor 1) belongs to the tumor necrosis factor superfamily, and is one of the major TNF-alpha receptors. TNFR1 plays an important role in mediating, in cytokine mediated signaling, positive regulation of the NF-Kb pathway, positive regulation of angiogenesis, and negative regulation of gene expression. The extracellular domain of TNFR1 is also released into the circulatory system as soluble TNFR1 (sTNFR1). In humans, the TNFR1 gene is located on chromosome 12. Anti-apoptotic protein BCL2-associated athanogene 4 (BAG4/SODD) and adaptor proteins TRADD and TRAF2 have been shown to interact with TNFR1, and thus play regulatory roles in the signal transduction mediated by the receptor. Germline mutations of the extracellular domains of TNFR1 were found to be associated with the autosomal domit periodic fever syndrome, and the impaired receptor clearance is thought to be a mechanism of the disease.
Specifications
Specifications
TNFR1
Monoclonal
1.44 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P19438, P22934, P25118, P50555
Tnfrsf1a
TNFR1 Fusion Protein Ag16112
150 μL
Primary
Human, Mouse, Porcine, Rat
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
2A6E3
Unconjugated
TNFRSF1A
CD120a, FPF, p55, p55 R, p60, TBP1, TBPI, TNF R, TNF R I, TNF R1, TNF R55, TNF Receptor 1, TNF Receptor I, TNF RI, TNFAR, TNFR I, TNFR1, TNFR55, TNFR60, TNFRSF1A
Mouse
Protein G
RUO
21937, 25625, 397020, 7132
-20°C