Learn More
Tocris Bioscience™ Tobramycin
Aminoglycoside antibiotic
Brand:  Tocris Bioscience™ 5390/50
63.93 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18712128
  • 72.00 € / 50mg
Explore more special offers
Description
Description
Tobramycin is an aminoglycoside antibiotic. Exhibits antibacterial activity against strains of Enterobacteriaceae, Pseudomonas and Staphylococcus aureus. Shown to protect lung epithelial cells in vitro against injury from cystic fibrosis bronchial secretions and H2O2.
Specifications
Specifications
50 mg
Tobramycin
467.52
Tobramycin