missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocrifluor T1117, Tocris Bioscience™
Click to view available options
Quantity:
100 μg
Description
Fluorescent form of AM 251 . AM 251 conjugated with 5-carboxytetramethylrhodamine (5-TAMRA) that fluoresces at 543 nm excitation (590 nm emission). Displays GPR55 binding activity.
View more information regarding Tocrifluor T1117 .
Specifications
Specifications
CAS | 1186195-59-4 |
Molecular Formula | C56H53Cl2N7O5 |
Synonym | N-(Piperidin-1-yl)-5-(4-(4-(3-(5-carboxamidotetramethylrhodaminyl)propyl))phenyl)-1-(2, 4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide |
InChI Key | RNAPRXIZPJDYPH-UHFFFAOYSA-N |
SMILES | CC1=C(N(N=C1C(=O)NN2CCCCC2)C3=C(C=C(C=C3)Cl)Cl)C4=CC=C(C=C4)C5=CC=C(C=C5)CCCNC(=O)C6=CC7=C(C=C6)C8(C9=C(C=C(C=C9)N(C)C)OC1=C8C=CC(=C1)N(C)C)OC7=O |
IUPAC Name | 5-[4-[4-[3-[[3',6'-bis(dimethylamino)-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carbonyl]amino]propyl]phenyl]phenyl]-1-(2,4-dichlorophenyl)-4-methyl-N-piperidin-1-ylpyrazole-3-carboxamide |
Molecular Weight (g/mol) | 974.984 |
PubChem CID | 57369426 |
Formula Weight | 974.97 |
Percent Purity | >95% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'