Learn More
Tocris Bioscience™ BDNF (human)
Activates TrkB and p75 receptors
Brand:  Tocris Bioscience™ 2837/10U
363.87 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18004612
  • 464.00 € / 10µg
Explore more special offers
Description
Description
BDNF (human) is a member of the neurotrophin growth factor family that binds and activates TrkB and p75 neurotrophin receptors. BDNF enhances the survival, growth and differentiation of neurons. BDNF expression is altered in neurodegenerative disorders such as Parkinson's and Alzheimer's disease. In an animal model of Huntington's disease, BDNF administration or overexpression rescues the disease phenotype.
Specifications
Specifications
0.95