Learn More
Tocris Bioscience™ (d(CH2)51,Tyr(Me)2,Arg8)-Vasopressin
Selective vasopressin V1A antagonist
Brand:  Tocris Bioscience™ 3377/1
159.36 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18055972
  • 201.00 € / 1mg
Explore more special offers
Description
Description
(d(CH2)51,Tyr(Me)2,Arg8)-Vasopressin is a selective vasopressin V1A receptor antagonist. Inhibits vasopressin and oxytocin-induced increases in intracellular calcium concentrations in vitro (IC50 values are 5 and 30 nM respectively). Exhibits potent and prolonged antivasopressor activity and induces anxiolytic-like effects in the dorsal, but not ventral, hippocampus in vivo.
Specifications
Specifications
0.95