Learn More
Tocris Bioscience™ Exendin-4
Potent GLP-1 receptor agonist
Brand:  Tocris Bioscience™ 1933/1
380.98 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18787186
  • 497.00 € / 1mg
Explore more special offers
Description
Description
Exendin-4 is a high affinity glucagon-like peptide 1 (GLP-1) receptor agonist (Kd = 136 pM); originally isolated from Heloderma suspectum venom. Exendin-4 potently induces cAMP formation without stimulating amylase release in pancreatic acini. Potentiates glucose-induced insulin secretion in isolated rat islets. Exendin-4 protects against glutamate-induced neurotoxicity, and in a mouse model of metabolic imblance it reduces neuroinflammation and enhances long-term-potentiation.