Learn More
Tocris Bioscience™ GIP (human)
Potent insulinotropic gut hormone; GIP agonist
Brand:  Tocris Bioscience™ 2084/1
Product Code. 18044312
  • 596.00 € / 1mg
Explore more special offers
Description
Description
GIP (human) is a potent insulinotropic hormone synthesized by duodenal K-cells. High affinity GIP receptor agonist (EC50 = 0.81 nM) that inhibits gastric acid secretion and stimulates pancreatic insulin release in response to glucose. Also affects lipid metabolism and displays mitogenic and antiapoptotic effects in pancreatic β-cells.
Specifications
Specifications
0.95
Store at -20°C