Learn More
Tocris Bioscience™ GLYX 13
NMDA receptor partial agonist; acts at the glycine site
Brand:  Tocris Bioscience™ 3406/1
181.50 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18034112
  • 225.00 € / 1mg
Explore more special offers
Description
Description
GLYX 13 is a NMDA receptor partial agonist that acts at the glycine site. Simultaneously acts as a promoter of the induction of long-term potentiation (LTP) and as a suppressor of long-term depression (LTD). Also promotes VGF and BDNF release and increases ERK/mTOR signaling . Exhibits nootropic, neuroprotective and antinociceptive activity, and enhances learning, memory and cognition in vivo. Brain penetrant.
Specifications
Specifications
0.95