Learn More
Tocris Bioscience™ N-Formyl-Met-Leu-Phe
Endogenous FPR1 agonist
Brand:  Tocris Bioscience™ 1921/1
102.70 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18073962
  • 128.00 € / 1mg
Explore more special offers
Description
Description
N-Formyl-Met-Leu-Phe is an endogenous chemotactic peptide and agonist for the formyl peptide receptor 1 (FPR1) (Ki = 38 nM). Stimulates aggregation of leukocytes.
Specifications
Specifications
0.95
Store at -20°C