Learn More
Tocris Bioscience™ Leupeptin hemisulfate
Inhibits trypsin-like/cysteine proteases
Brand:  Tocris Bioscience™ 1167/25
173.65 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18792494
  • 227.00 € / 25mg
Explore more special offers
Description
Description
Leupeptin hemisulfate is a reversible inhibitor of trypsin-like and cysteine proteases such as calpain. Shown to inhibit activation-induced programmed cell death.
Specifications
Specifications
0.95
Store at -20°C
25 mg