Learn More
Tocris Bioscience™ MCH (human, mouse, rat)
Potent endogenous MCH agonist
Brand:  Tocris Bioscience™ 3806/100U
102.70 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18075782
  • 130.00 € / 100µg
Explore more special offers
Description
Description
MCH (human, mouse, rat) is a potent endogenous agonist at melanin-concentration hormone (MCH) receptors (IC50 values are 0.3 and 1.5 nM and EC50 values are 3.9 and 0.1 nM at MCH1 and MCH2 receptors respectively). Increases food intake in vivo.