Learn More
Tocris Bioscience™ Neurokinin B (human, porcine)
Endogenous tachykinin peptide
Brand: Tocris Bioscience™ 1582/1
206.28 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Neurokinin B (human, porcine) is an endogenous tachykinin agonist peptide that shows preference for the NK3 receptor (EC50 = 1 nM).Specifications
95% | |
Store at -20°C | |
1 mg |