Learn More
Tocris Bioscience™ Tocriscreen Antiviral Library
Antiviral compound library for high throughput screening, chemical biology applications and drug repurposing; contains 240 bioactive compounds (100 μL 10 mM DMSO solutions)
Brand:  Tocris Bioscience™ 7350/1 SET
2513.11 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18779346
  • 3240.00 € / 1 set
Explore more special offers
Description
Description
Tocriscreen Antiviral Library is an Antiviral compound library composed of 240 compounds (100 μL 10 mM DMSO). Contains a diverse collection of compounds to investigate the entire viral lifecyle as well as cytokine storm. Suitable for high throughput screening, high content screening, chemical biology applications and drug repurposing studies. Includes high-interest compounds such as remdesivir (Cat. No. 7226) as well as innovative products like MPro N3 (Cat. No. 7230).
If this library does not suit your needs, please submit your requirements through our Tocriscreen PRO custom compound library service .
Specifications
Specifications
1 Set
Screening Libraries