Learn More
Tocris Bioscience™ Tocriscreen GPCR Compound Library
A bioactive compound library of 240 GPCR compounds (100 μL 10 mM DMSO solutions) for high-throughput screening, high-content screening, target validation and assay development
Brand:  Tocris Bioscience™ 8100/1 Set
2540.75 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18701854
  • 3180.00 € / 1 set
Explore more special offers
Description
Description
Tocriscreen GPCR Compound Library is a library of 240 compounds (100 μl, 10 mM) for GPCR research. Contains compounds that target >30 types of GPCR, including peptide receptors, 5-HT receptors, chemokine receptors, adenosine receptors, adrenergic receptors and more. Adapted for popular techniques in drug discovery and cell biology research, including high throughput screening, high content screening, target validation and assay development.If this library does not suit your needs, please submit your requirements through our Tocriscreen PRO custom compound library service .
Specifications
Specifications
1 Set
Screening Libraries