missing translation for 'onlineSavingsMsg'
Learn More

Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals

Product Code. 11374257
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Toluene diisocyanate is utilized in polyurethane foams and tank trucks. It is also used in concrete sealers for aircraft, sealers and coatings in floor. It is employed as a crosslinking agent for nylon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-84-9
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00002011
InChI Key DVKJHBMWWAPEIU-UHFFFAOYSA-N
Synonym 2,4-toluene diisocyanate, 2,4-diisocyanatotoluene, tolylene-2,4-diisocyanate, toluene-2,4-diisocyanate, 2,4-tolylene diisocyanate, toluene 2,4-diisocyanate, 4-methyl-m-phenylene diisocyanate, tolylene diisocyanate, 2,4-tdi, toluylene-2,4-diisocyanate
PubChem CID 11443
ChEBI CHEBI:53556
IUPAC Name 2,4-diisocyanato-1-methylbenzene
SMILES CC1=C(C=C(C=C1)N=C=O)N=C=O

Specifications

Melting Point 19.5°C to 21.5°C
Color Colorless
Density 1.225 g/mL
Boiling Point 251°C
Flash Point 132° (270°F)
Refractive Index 1.567
Quantity 1 kg
UN Number UN2078
Beilstein 744602
Sensitivity Moisture sensitive
Merck Index 14,9530
Solubility Information Miscible with ether,acetone,benzene,carbontetrachloride and chlorobenzene.
Formula Weight 174.16
Percent Purity 80%
Grade Technical
Physical Form Liquid
Assay Remainder 2,6-diisocyanate
Chemical Name or Material Toluene-2,4-diisocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.