missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene-4-Sulfonic Acid, Extra Pure, SLR, Fisher Chemical™
51.10 €
Chemical Identifiers
CAS | 6192-52-5 |
---|---|
Molecular Formula | C7H10O4S |
Molecular Weight (g/mol) | 190.213 |
MDL Number | 142137 |
InChI Key | KJIFKLIQANRMOU-UHFFFAOYSA-N |
Synonym | p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate |
PubChem CID | 521998 |
IUPAC Name | 4-methylbenzenesulfonic acid;hydrate |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)O.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10686462
|
Fisher Chemical
T/2500/53 |
500 g | N/A | ||||||
Description
Chemical Identifiers
6192-52-5 | |
190.213 | |
KJIFKLIQANRMOU-UHFFFAOYSA-N | |
521998 | |
CC1=CC=C(C=C1)S(=O)(=O)O.O |
C7H10O4S | |
142137 | |
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate | |
4-methylbenzenesulfonic acid;hydrate |
Specifications
100°C | |
White | |
Solid | |
142137 | |
KJIFKLIQANRMOU-UHFFFAOYSA-N | |
4-methylbenzenesulfonic acid;hydrate | |
521998 |
6192-52-5 | |
Plastic Powder Jar | |
C7H10O4S | |
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate | |
CC1=CC=C(C=C1)S(=O)(=O)O.O | |
190.213 | |
190.22g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- p-Toluenesulfonic acid monohydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.