missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, 99+%, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Container
Plastic-coated Glass Bottle
Description
Toluene, 99+%, Extra Pure, SLR

Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Boiling Point | 111°C |
Quantity | 500 mL |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 29mbar at 20°C |
Viscosity | 0.6 mPaS at 20°C |
Formula Weight | 92.14g/mol |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2527234 | Certificate of Analysis | 17/02/2025 | T/2200/PB17,T/2200/21,T/2200/FP21,T/2200/25,T/2200/27,T/2200/17,T/2200/08,T/2200/21S |
Safety and Handling



Product Identifier
- Toluene
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Toluene, 99+%, Extra Pure, SLR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction