missing translation for 'onlineSavingsMsg'
Learn More
Toluene, 99+%, extra pure, Thermo Scientific Chemicals Product Code.: 10702991

Toluene, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10702991
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10702991

Brand: Thermo Scientific Acros 177160010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95.0°C
Density 0.8660g/mL
Boiling Point 111.0°C
Quantity 1 L
Flash Point 4°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4968
Assay Percent Range 99+%
Residue after Evaporation 0.003% max.
Specific Gravity 0.866
Linear Formula C6H5CH3
Vapor Pressure Vapor Pressure: 29mbar at 20°C
Viscosity 0.6 mPa.s (20°C)
Beilstein 05, 280
Water 0.05% max. (K.F.)
Merck Index 15, 9688
Solubility Information Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
Formula Weight 92.14
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Toluene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2533994 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number177160025,177160010,177160100,177160050,177160250
Lot Number2411693 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number177160025,177160010,177160100,177160050,177160250
Lot Number2421395 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number177160025,177160010,177160100,177160050,177160250
Lot Number2401187 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number177160025,177160010,177160100,177160050,177160250
Lot Number2404618 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number177160025,177160010,177160100,177160050,177160250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Toluene, 99+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.