Learn More
Toluene, 99+%, for spectroscopy, Thermo Scientific Chemicals
98.10 € - 189.00 €
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10114330
|
Thermo Scientific Acros
167900010 |
1 L | Glass Bottle |
98.10 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10174370
|
Thermo Scientific Acros
167900025 |
2.5 L | Glass Bottle |
189.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopyChemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Specifications
(1 cm cell vs water), 0.01 max. at 350nm, 0.02 max. at 335nm, 0.07 max. at 310nm, 0.1 max. at 300nm, 0.4 max. at 288nm, 1 max. at 285nm | |
0.8660g/mL | |
99+% | |
C7H8 | |
MFCD00008512 | |
05, 280 | |
15, 9688 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 | |
Spectroscopy |
108-88-3 | |
Liquid | |
0.001% max. | |
C6H5CH3 | |
Vapor Pressure: 29mbar at 20°C | |
0.03% max. | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
≥99.0% | |
Toluene, 99+% |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.
Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
Signal Word: DANGER!
missing translation for 'einecsNumber' : 203-625-9
missing translation for 'rtecsNumber' : XS5250000
missing translation for 'tsca' : TSCA
- Toluene
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
RUO – Research Use Only