missing translation for 'onlineSavingsMsg'
Learn More
Toluene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals Product Code.: 11374807

Toluene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 11374807
250 mL
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11374807

Brand: Thermo Scientific Alfa Aesar 041841.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Toluene is widely used as an organic solvent in various industries like paints, pharmaceuticals, dyes, agrochemicals and polymers. It is used as an additive in gasoline and improves octane ratings, as a fullerene indicator, in paint thinners and silicone sealants, as a coolant in sodium cold traps used in nuclear reactor system loops and as a fuel for both two-stroke and four-stroke engines. It is also used as a raw material in the production of benzene, benzaldehyde, benzoic acid, toluene diisocyanate, benzyl-tert-butyl peroxide, xylene and many other chemical products. It plays an important role in biochemistry experiments to break open red blood cells in order to extract haemoglobin as well as used in the removal of cocaine from coca leaves to make coca-cola syrup. Further, it used as a standard material for chromatography and as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point °C to 93°C
Density 0.867 g/mL
Boiling Point 111°C
Quantity 250 mL
Flash Point 4°C (40°F)
Physical Form Liquid
Refractive Index 1.4967
Assay Percent Range 99.8%
Beilstein 635760
Solubility Information Miscible with ethanol,alcohol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water.
Formula Weight 92.14
Chemical Name or Material Toluene, anhydrous
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene, anhydrous
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Toluene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Toluene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.