missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Description
C7H8, CAS Number-108-88-3, monomethyl benzene, phenylmethane, methylbenzene, methylbenzol, benzene, methyl, methacide, antisal 1a, tolu-sol, toluen, toluol, 2.5L, 111 deg.C, CHEBI:17578, Colorless, 92.14g/mol, YXFVVABEGXRONW-UHFFFAOYSA-N, toluene, -95 deg.C, 8512, 92.141, Amber glass bottle, Liquid

Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Boiling Point | 111°C |
Quantity | 5 L |
Packaging | Metal Can |
Physical Form | Liquid |
Vapor Pressure | 29mbar at 20°C |
Viscosity | 0.6 mPaS at 20°C |
Formula Weight | 92.14g/mol |
Safety and Handling



Product Identifier
- Toluene
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Toluene, Certified AR for Analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction