missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Toluene CHROMASOLV™

Product Code. 15965470
Click to view available options
Quantity:
7 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Toluene CHROMASOLV™ for analysis of dioxins, furans and PCB, ≥99.7%.

  • High purity chromatography solvents.
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.

Specifications

pH No data available
Melting Point -95°C
CAS 108-88-3
Color Colorless
CAS Max % 100.00%
Density 0.867 g/cm3 (20°C)
Boiling Point 110.6°C
Quantity 7 L
Flash Point 7.1 %(V)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV™
Molecular Formula C7H8
Linear Formula C6H5CH3
MDL Number MFCD00008512
Vapor Pressure 38 hPa (25°C)
UN Number UN1294
Viscosity No data available
Beilstein 635760
Synonym Methylbenzene, phenylmethane
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1
IUPAC Name toluene
Molecular Weight (g/mol) 92.14
Percent Purity ≥99.70%
Grade GC
Chemical Name or Material Toluene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.