missing translation for 'onlineSavingsMsg'
Learn More

α-Toluenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10581231
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1939-99-7
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.64
MDL Number MFCD00007455
InChI Key OAHKWDDSKCRNFE-UHFFFAOYSA-N
Synonym alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride
PubChem CID 74740
IUPAC Name phenylmethanesulfonyl chloride
SMILES C1=CC=C(C=C1)CS(=O)(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 90°C to 94°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula C6H5CH2SO2Cl
Refractive Index 1.6240 to 1.626
Quantity 25 g
Beilstein 11, 116
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in ethanol and ether
Formula Weight 190.64
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material α-Toluenesulfonyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • a-Toluenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
α-Toluenesulfonyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.