Learn More
MP Biomedicals Top of CryoPrep Adapter
FastPrep-24 replacement part.
Brand:  MP Biomedicals 116002528T
Product Code. 15752432
  • 483.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Adapter type: Cryogenic
  • Sample throughput: Medium
  • Tube compatibility: 2 mL
  • For FastPrep-24
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 mL
Replacement Top Component
FastPrep-24™
Sample throughput: Medium
Holding plate to secure tubes and adapter
24 x 2 mL Tubes