Learn More
Fisherbrand™ Top-Line Pipette Tips
Eliminate sample contamination and prevent sample loss. . Fisherbrand™ Top-Line Pipette Tips provide precision pipetting with conical tip and secure fit.
9.15 € - 51.20 €
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO Standard 8655, EU-Directive 98/79/EG, competent authority code DE/CA90, EN 552/ISO 11137 and DIN EN 556, EU Directive 2002/95/EC, ISO Standard 14000 and ISO Standard 14001, ISO Standard 10993, DIN Standard 12600
Tip Style Universal
For Use With (Equipment) Universal Fit
Graduated Yes
View More Specs
Products 11
Product Code Brand Volume (Metric) Length (Metric) Sterility Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Length (Metric) Sterility Color Price Quantity & Availability  
10182742
View Documents
Fisherbrand™
FI1-000-96-0
0.1 to 10 μL 34.1mm Non-sterile Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10346052
View Documents
Fisherbrand™
FI1-050-60-0
0.5 to 20 μL 45.7mm Non-sterile Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10739444
View Documents
Fisherbrand™
FI1-110-96-0
1 to 200 μL 50.8mm Non-sterile Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10739254
missing translation for 'viewDocuments'
Fisherbrand™
FI1-110-81-1
1 to 200 μL 50.8mm Non-sterile Yellow This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10233932
missing translation for 'viewDocuments'
Fisherbrand™
FI1-001-96-0
0.1 to 10 μL 34.1mm Sterile Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10458072
missing translation for 'viewDocuments'
Fisherbrand™
FI1-111-96-0
1 to 200 μL 50.8mm Sterile Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10142172
missing translation for 'viewDocuments'
Fisherbrand™
FI1-110-96-1
1 to 200 μL 50.8mm Sterile Yellow This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10532543
missing translation for 'viewDocuments'
Fisherbrand™
FI1-050-96-0
0.5 to 20 μL 45.7mm Non-sterile Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10542543
missing translation for 'viewDocuments'
Fisherbrand™
FI1-051-96-0
0.5 to 20 μL 45.7mm Sterile Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10509862
missing translation for 'viewDocuments'
Fisherbrand™
FI1-103-81-0
5 to 300 μL 55mm Non-sterile Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10396142
missing translation for 'viewDocuments'
Fisherbrand™
FI1-103-96-0
5 to 300 μL 55mm Non-sterile Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Optimized cone geometry for a secure fit
  • Graduation marks on tip at standard volumes
  • Conical tip opening prevents drop formation at the end of the tip
  • Tips made of autoclavable, non-wettable, virgin polypropylene
  • Sterile packs available (beta irradiation)
Specifications
Specifications
Autoclavable
Universal
Yes
Virgin Polypropylene
ISO Standard 8655, EU-Directive 98/79/EG, competent authority code DE/CA90, EN 552/ISO 11137 and DIN EN 556, EU Directive 2002/95/EC, ISO Standard 14000 and ISO Standard 14001, ISO Standard 10993, DIN Standard 12600
Universal Fit
Top-Line