missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOP10 Electrocomp™ Kit

Product Code. p-4924253
Click to view available options
Quantity:
5 x 100 μL
10 x 100 μL
30 x 100 μL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10398162

Brand: Invitrogen™ C66455

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TOP10 Electrocomp E. coli cells are provided at a transformation efficiency of 1 x 1010 cfu/μg supercoiled DNA and are ideal for high-efficiency cloning and plasmid propagation.

TOP10 Electrocomp E. coli cells are provided at a transformation efficiency of 1 x 1010 cfu/μg supercoiled DNA and are ideal for high-efficiency cloning and plasmid propagation. This strain is used in wide range of applications and allows stable replication of high-copy number plasmids and maintenance of large plasmids. 

The TOP10 genotype is similar to the DH10B strain and features:
hsdR for efficient transformation of unmethylated DNA from PCR amplifications
mcrA for efficient transformation of methylated DNA from genomic preparations
lacZΔM15 for blue/white color screening of recombinant clones
endA1 for cleaner preparations of DNA and better results in downstream applications due to the elimination of non-specific digestion by Endonuclease I
recA1 for reduced occurrence of non-specific recombination in cloned DNA

Notes
Please see the product manual for reaction scale recommendations. A high-voltage electroporation apparatus is required.

Genotype
F- mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK rpsL endA1 nupG

Find the strain and format that fits your needs
• TOP10 is available in chemically competent and electrocompetent cell formats.
• Choose MultiShot competent cells for high-throughput cloning applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Electrocompetent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level High Efficiency (>1 x 1010 cfu/μg)
Content And Storage • 5 x 100μL TOP10 Electrocomp E. coli
• 50 μL pUC19 vector (10 pg/μL)
Store at –80°C.

• S.O.C. Medium (15 mL)
Store at room temperature.

Antibiotic Resistance Bacterial Yes (Streptomycin)
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacΔM15)
High-throughput Compatibility Low
Plasmid May be used for plasmids >20 kb
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Format Tube
Product Line Electrocomp
Quantity 5 x 100 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.