Learn More
Topiramate, MedChemExpress
MedChemExpress Topiramate (McN 4853) is a broad-spectrum antiepileptic agent. Topiramate is a GluR5 receptor antagonist. Topiramate produces its antiepileptic effects through enhancement of GABAergic activity, inhibition of kainate/AMPA receptors, inhibition of voltage-sensitive sodium and calcium channels, increases in potassium conductance, and inhibition of carbonic anhydrase.
Brand:  Medchem Express HY-B0122
Encompass_Preferred
 View more versions of this product
Product Code. 16458116
  • 137.70 € / 100mg
Explore more special offers
Description
Description
Topiramate (McN 4853) is a broad-spectrum antiepileptic agent. Topiramate is a GluR5 receptor antagonist. Topiramate produces its antiepileptic effects through enhancement of GABAergic activity, inhibition of kainate/AMPA receptors, inhibition of voltage-sensitive sodium and calcium channels, increases in potassium conductance, and inhibition of carbonic anhydrase.
Specifications
Specifications
Topiramate
White
C12H21NO8S
McN 4853 RWJ 17021
NS(OC[C@]1(OC(C)(C)O2)[C@@H]2[C@H](OC(C)(C)O3)[C@H]3CO1)(=O)=O
339.36
Research
Neuroscience-Neuromodulation
97240-79-4
Research
100 mg
DMSO : ≥ 100 mg/mL (294.67 mM) ∣H2O : 4 mg/mL (11.79 mM; Need ultrasonic)
339.36
98.0%
Solid