Learn More
3M™ Tora™ Safety Glasses
Click to view available options
Lens Tint: Bronze
Description
- Safety glasses with a wraparound lens design offering excellent peripheral vision and lateral side protection
- Anti-scratch and anti-fog coating for increased durability and improved vision
- Extremely lightweight at only 22 g
- Adjustable neck cord for increased comfort and convenient storage
Specifications
Specifications
Certifications/Compliance | CE Approved, EN 166:2001 |
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Bronze |
Product Type | Safety glasses |
Lens Material | Polycarbonate |