missing translation for 'onlineSavingsMsg'
Learn More

Tosylmethyl isocyanide, 98%, Thermo Scientific Chemicals

Product Code. 10447582
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36635-61-7
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
MDL Number MFCD00000005
InChI Key CFOAUYCPAUGDFF-UHFFFAOYSA-N
Synonym tosylmethyl isocyanide, tosmic, p-toluenesulfonylmethyl isocyanide, 4-toluenesulphonylmethyl isocyanide, 4-toluenesulfonylmethyl isocyanide, tosylmethylisocyanide, 1-isocyanomethylsulfonyl-4-methylbenzene, p-tolylsulfonyl methyl isocyanide, unii-c35fd6olh8, 1-isocyanomethyl sulfonyl-4-methylbenzene
PubChem CID 161915
IUPAC Name 1-(isocyanomethylsulfonyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)C[N+]#[C-]

Specifications

Melting Point 111.0°C to 113.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H4SO2CH2NC
Quantity 5 g
Fieser 04,514; 05,684; 06,600; 07,377; 08,493; 10,409; 11,539; 12,511; 13,313; 17,20
Merck Index 15, 9715
Solubility Information Solubility in water: insoluble
Formula Weight 195.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tosylmethyl isocyanide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tosylmethyl isocyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.