Learn More
WTW™ Total Nitrogen Test Kits
Obtain precision results using the CombiCheck Total Nitrogen NTotal cell ready-made test kit for the determination of total nitrogen in waste water, drinking water, and surface water.
182.00 € - 197.00 €
Specifications
For Use With (Application) For waste water, drinking water, surface water
Test Method Photometric
View More Specs
Products 3
Product Code Brand Range Price Quantity & Availability  
Product Code Brand Range Price Quantity & Availability  
11754725
View Documents
WTW™
250494
10 to 150 mg/L
182.00 €
0.15kg
10201595
View Documents
WTW™
252018
0.5 to 15mg/L
184.00 €
Each
11751073
View Documents
WTW™
250358
0.5 to 15 mg/L NTotal (120°C/248°F, 1 h)
197.00 €
0.25kg
Description
Description
  • Variety of instrumentation for testing nitrogen in water
  • For waste water, drinking water, surface water
  • Nitrospectral Method
  • Photometric Test Methods
  • CombiCheck Total Nitrogen NTotal cell test kit
  • Safe determinations of COD and NTotal in water
Specifications
Specifications
For waste water, drinking water, surface water
Photometric
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
WTW™ Total Nitrogen Test Kits