Learn More
Total Organic Carbon (TOC), standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals
Brand:  Thermo Scientific Alfa Aesar 042562.AE
 View more versions of this product
Product Code. 11329168
  • 141.71 € / 100mL
Explore more special offers
Description
Description
Total Organic Carbon (TOC), is used as a standard solution during the measurement of chemical analysis such as detecting contaminants in drinking water and cooling water. It is also used for analysis of water to be used for semiconductor manufacturing and pharmaceutical use.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Total Organic Carbon
100 mL
KHC8H4O4 in H2O
Characteristic
Liquid
1000μg/mL
Colorless
MFCD00133992
Miscible with water