Learn More
Diversey™ Touchless Dispenser
Touchless dispenser for use with ClearKlens IPA.
Brand:  Diversey™ 4400420
Product Code. 17613105
  • 549.00 € / Each
Explore more special offers
Description
Description
  • Pump technology proven in continuous use
  • Selection of materials according to hygienic viewpoints
  • Touchless operation optimizes hand hygiene
  • Simple pump removal from the front
  • Battery level indicator
  • Suitable for Sterile ClearKlens™ IPA bottles
  • Lockable cover available as an accessory
Body
  • High hygienic standard through anodized aluminum surfaces
  • Stress resistant construction optimized for continuous use
  • Exchangeable face shield for individual product labelling
Pump and dosage
  • IMP Touchless stainless steel pump, angled stainless steel suction tube, autoclavable
  • Dosage amount: approx. 1.5 mL/push, reducible to approx. 0.5 mL/push
Filling: Designed to be used with ClearKlens IPA (70% IPA, 30% WFI)
Specifications
Specifications
Touch-free Dispenser
ClearKlens IPA