Learn More
Ansell™ TouchNTuff™ 83-300 Gloves
BIOPHARMA
Product Code. 15975011
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Ansell™ TouchNTuff™ 83-300 is an ultra-soft polyisoprene glove that offers the performance of natural rubber latex without the risk of latex sensitization.
  • Compatible with Class 100 (ISO 5) / Grade A Cleanroom Environments
  • Added comfort with second skin feel
  • Latex-free polyisoprene performance
  • Prevents Type I allergies
  • Extended cuff
  • Cuff style is beaded with Surefit™ Technology
  • Freeedom from holes 0.65 AQL
  • Double glove recommended
  • Internal glove surface is coated
  • Anatomic with curved fingers
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D2577, CE Marked CE0493
Description Smooth external glove surface
Color White
Material Polyisoprene
Product Type Disposable Gloves
Size 7.5
Thickness (English) 0.25 mil
Thickness (Metric) 0.17 mm
EN 421 Yes
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KPT
EN ISO 374 - 5 Yes
Glove Cuff Style Beaded
Packaging Type Polybag
Quantity 20 pairs
Length (English) Cuff 11.6 in.
Length (Metric) Cuff 295 mm