Learn More
Ansell TouchNTuff™ 93-700 Series Green Nitrile Gloves Product Code.: 13429028
Add to basket
Ansell TouchNTuff™ 93-700 Series Green Nitrile Gloves
BIOPHARMA
Product Code. 13429028 Shop All Ansell Products
10
Click to view available options
Size:
7
8
9
10
Product Code. 13429028
Brand: Ansell 93700100
Add to basket
Add to basket
Delivers the highest protection for a wide range of chemicals than any other single use nitrile glove. Manufactured from nitrile, these gloves offer three times the puncture resistance of standard latex gloves.
  • Provides the wearer with a high level of comfort and increased protection in the forearm with longer cuff.
  • Free of natural rubber latex proteins, the glove may be worn with confidence by latex sensitive (Type I) wearers.
  • 93-700: Each glove is post cleaned, single pair packed and sterilized.
TRUSTED_SUSTAINABILITY
Specifications
Cleanroom Class Class 100 (ISO 5)
Exterior Finish Fully Textured
Product Type Cleanroom Gloves
Size 10
Thickness (Metric) 0.135 mm
For Use With (Application) For blending of liquids, filtration processes, vaccines injectables operations etc.
Model 9370010
Quantity 200/Pk.
Resistant To Splash and high puncture resistance
Series 93-700
Product Content Correction
Product Title
Ansell TouchNTuff™ 93-700 Series Green Nitrile Gloves >