Learn More
Ansell™ TouchNTuff™ Disposable Blue Nitrile Gloves
Innovative nitrile compound offers outstanding chemical and mechanical properties, comfort and dexterity. Ansell TouchNTuff™ Disposable Blue Nitrile Gloves are lighter and cooler, yet safer and more durable.
22.70 €
Specifications
Certifications/Compliance EN420, EN374/1,2,3, ASTM D 6319, FDA 21 CFR 177-2600,Arrete 09/11/04, European Regulation 1935/2004,German Recommendation for Food Commodities.
Color Blue
Disposable Single-use
Laboratory/Cleanroom Use Yes
Latex Free Yes
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11726843
View Documents
Ansell™
92670100
10
22.70 €
Pack of 100
In Stock
to see stock.
Add to basket
11766803
View Documents
Ansell™
92670070
7
22.70 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
11776803
View Documents
Ansell™
92670080
8
22.70 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
11786803
View Documents
Ansell™
92670090
9
22.70 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Three times the puncture resistance of latex or PVC disposable gloves
  • 100% nitrile; no waxes, silicone or plasticizers
  • Powder-free
  • No risk of contaminating food with corn starch used to powder gloves
  • Textured finish on fingertips
  • Improved grip
  • Rolled cuff
  • Provides added protection at wrist and ensures a secure fit
Specifications
Specifications
EN420, EN374/1,2,3, ASTM D 6319, FDA 21 CFR 177-2600,Arrete 09/11/04, European Regulation 1935/2004,German Recommendation for Food Commodities.
Single-use
Yes
Powder Free
Rolled
240 mm
Blue
Yes
Nitrile
0.11 mm
92-670
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Ansell™ TouchNTuff™ Disposable Blue Nitrile Gloves