missing translation for 'onlineSavingsMsg'
Learn More

TPEN, Tocris Bioscience™

Product Code. 18085592
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cell-permeable Zn2+ chelator; also inhibits RNA binding protein Lin28

Heavy metal chelator. Reacts with both Zn-proteome and Zn-metallothionein (MT) in LLC-PK1 cells; acts as an intracellular chelator of proteomic Zn2+. Activity decreases intracellular zinc levels, and induces apoptosis in HeLa and cultured human retinal pigment epithelium (RPE) cells. Also inhibits RNA binding protein Lin28 (IC50 = 2.5 μM). Cell permeable.

Specifications

CAS 16858-02-9
Molecular Formula C26H28N6
MDL Number MFCD00036918
Quantity 100 mg
Synonym tpen, n,n,n',n'-tetrakis 2-pyridylmethyl ethylenediamine, n1,n1,n2,n2-tetrakis pyridin-2-ylmethyl ethane-1,2-diamine, unii-r9ptu1u29i, n,n,n',n'-tetrakis 2-pyridylmethyl ethane-1,2-diamine, r9ptu1u29i, n,n,n',n'-tetrakis 2-pyridyl-methyl ethylenediamine, 1,2-ethanediamine, n,n,n',n'-tetrakis 2-pyridinylmethyl-, oc-6-11, 1,2-ethanediamine,n,n,n',n'-tetrakis 2-pyridinylmethyl, 1,2-ethanediamine, n,n,n',n'-tetrakis 2-pyridinylmethyl
InChI Key CVRXLMUYFMERMJ-UHFFFAOYSA-N
SMILES C(CN(CC1=CC=CC=N1)CC1=CC=CC=N1)N(CC1=CC=CC=N1)CC1=CC=CC=N1
IUPAC Name (2-{bis[(pyridin-2-yl)methyl]amino}ethyl)bis[(pyridin-2-yl)methyl]amine
Molecular Weight (g/mol) 424.55
PubChem CID 5519
Formula Weight 424.54
Percent Purity >98%
Chemical Name or Material TPEN
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.