missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ (TPX™) PMP Graduated Conical Measures

Facilitates easier pouring while not significantly decreasing measurement accuracy. Kartell™ Graduated Conical Measures provide the ability to mix solutions within the measure itself.

Brand:  Kartell™ 0142200

 View more versions of this product

Product Code. 10225141

  • 123.00 € / Pack of 12
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Can withstand temperatures up to 170°C
  • Wide base for stability
  • Suitable for applications with food
Specifications

Specifications

100 mL
2 mL
Cylindrical
Wide
Graduated Measuring Cylinder
12Pack
170°C
Foodstuff
117 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.