Learn More
Thermo Scientific™ TR-FRET Coregulator Buffer C
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Brand: Thermo Scientific™ PV4419
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.

Specifications
TR-FRET Coregulator Buffer C, 25mL, Liquid, 8 pH Range, Store at 4°C, Proprietary Buffer, Water Soluble, Used For Pharma & Biopharma, Target & Lead Identification & Validation | |
25mL, 25 mL | |
TR-FRET Coregulator Buffer C may be shipped frozen or at room temperature. Store at 4°C. |
Liquid | |
LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2944429B | Certificate of Analysis | 21/02/2025 | PV4419 |
2944429A | Certificate of Analysis | 21/02/2025 | PV4419 |
2894656A | Certificate of Analysis | 23/02/2024 | PV4419 |
2807801B | Certificate of Analysis | 01/02/2024 | PV4419 |
2807801A | Certificate of Analysis | 07/11/2023 | PV4419 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.