Learn More
Trace Metals Standards For Environmental Analysis, Set of 3, SPEX CertiPrep™
Trace Metals Ground and Wastewater Pollution Control Check Standards with Mercury for AA & ICP, 125 mL, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
Brand:  SPEX Certiprep TM-SET
Product Code. 15678335
  • 471.00 € / 1 set
Explore more special offers
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
  • Trace Metals
  • 125 mL volume
  • Includes Mercury
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Specifications
Trace Metals
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Environmental
Set of 3 Groundwater and Wastewater Pollution Control Check Standard
3 x 125 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product intended for laboratory use only